Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | 2 | 1226 | NP_005348.2 |
XM_005258937.3 | 2 | 1221 | XP_005258994.1 | |
XM_005258938.4 | 2 | 212 | XP_005258995.1 | |
XM_005258939.3 | 3 | 250 | XP_005258996.2 | |
XM_005258940.3 | 2 | 151 | XP_005258997.1 | |
XM_006723218.2 | 2 | 351 | XP_006723281.1 | |
XM_017026810.1 | 2 | 91 | XP_016882299.1 | |
AK292482.1 | 2 | 1206 | ||
BC070041.1 | 2 | 1226 | AAH70041.1 | |
U40002.1 | 2 | 1226 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | LIPE-AS1 LOC101930071 LIPE CNFN |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | LOC100505585 CD79A LIPE ERF DEDD2 MIR4323 LOC390937 CNFN MEGF8 ZNF526 ZNF574 PAFAH1B3 LOC100505622 ATP1A3 LIPE-AS1 LOC101930071 RPS19 GSK3A POU2F2 MIR8077 MIR6797 PRR19 RABAC1 ARHGEF1 TMEM145 GRIK5 CIC |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | ZNF526 PAFAH1B3 CXCL17 CEACAM8 LIPE-AS1 LOC101930071 LIPE ERF GSK3A POU2F2 DEDD2 MIR8077 CEACAM1 PRR19 TMEM145 CNFN CIC MEGF8 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | LOC100505585 CEACAM4 CD79A LIPE ERF DEDD2 MIR4323 CEACAM3 LOC390937 CNFN MEGF8 ZNF526 ZNF574 PAFAH1B3 LOC100505622 ATP1A3 LIPE-AS1 LOC101930071 RPS19 GSK3A LYPD4 POU2F2 CEACAM6 CEACAM5 MIR8077 MIR6797 DMRTC2 CEACAM7 PRR19 RABAC1 ARHGEF1 TMEM145 GRIK5 CIC |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | PSG10P PAFAH1B3 CXCL17 CEACAM8 LIPE-AS1 LOC101930071 PSG11 PSG1 LIPE ERF PSG7 GSK3A PSG3 MIR8077 CEACAM1 PSG6 PSG2 LOC100289650 PSG8 PRR19 TMEM145 CNFN CIC MEGF8 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |