Assay Details
Target Gene Details
Entrez Gene ID: | 4750 |
Gene Name: | NIMA related kinase 1 |
Gene Aliases: |
NY-REN-55, SRPS2, SRPS2A, SRTD6 |
Location: |
Chr.4:169393270-169612629 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEK1 | NM_001199397.1 | 34 | 4240 | NP_001186326.1 |
NM_001199398.1 | 33 | 4108 | NP_001186327.1 | |
NM_001199399.1 | 31 | 3949 | NP_001186328.1 | |
NM_001199400.1 | 31 | 4102 | NP_001186329.1 | |
NM_012224.2 | 32 | 4234 | NP_036356.1 | |
XM_006714228.1 | 33 | 4065 | XP_006714291.1 | |
XM_011532003.1 | 33 | 4158 | XP_011530305.1 | |
XM_011532004.1 | 32 | 4026 | XP_011530306.1 | |
XM_017008249.1 | 33 | 4144 | XP_016863738.1 | |
XM_017008250.1 | 32 | 4262 | XP_016863739.1 | |
XM_017008251.1 | 32 | 4060 | XP_016863740.1 | |
XM_017008252.1 | 30 | 4080 | XP_016863741.1 | |
XM_017008253.1 | 33 | 4090 | XP_016863742.1 | |
XM_017008254.1 | 26 | 3200 | XP_016863743.1 | |
AB067488.1 | 32 | 4070 | ||
AF155113.1 | 16 | 2118 | AAD42879.1 | |
AK025658.1 | 7 | 988 | BAB15207.1 | |
AL050385.1 | 33 | 4064 | CAI46225.1 | |
BC037790.2 | 6 | 900 | AAH37790.1 | |
BC114491.1 | 31 | 3709 | ||
CD358162.1 | 2 | 239 | ||
CR933642.1 | 34 | 4198 | CAI45943.1 | |
CR936672.1 | 31 | 4060 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |