Assay Details
Target Gene Details
Entrez Gene ID: | 653319 |
Gene Name: | KIAA0895 like |
Gene Aliases: |
- |
Location: |
Chr.16:67175602-67183980 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KIAA0895L | NM_001040715.1 | NP_001035805.1 | ||
AK092303.1 | BAC03856.1 | |||
AK094787.1 | ||||
AK096855.1 | ||||
AL834156.1 | CAD38862.1 | |||
BC007594.1 | AAH07594.1 |
Target Gene Details
Entrez Gene ID: | 8996 |
Gene Name: | nucleolar protein 3 |
Gene Aliases: |
ARC, FCM, MYP, NOP, NOP30 |
Location: |
Chr.16:67170497-67175737 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOL3 | NM_001185057.2 | 4 | 1360 | NP_001171986.1 |
NM_001276307.1 | 6 | 1394 | NP_001263236.1 | |
NM_001276309.1 | 4 | 1254 | NP_001263238.1 | |
NM_001276311.1 | 4 | 1246 | NP_001263240.1 | |
NM_001276312.1 | 6 | 1745 | NP_001263241.1 | |
NM_001276319.1 | 6 | 1745 | NP_001263248.1 | |
NM_003946.6 | 4 | 1370 | NP_003937.1 | |
XM_005256217.2 | 4 | 1997 | XP_005256274.1 | |
XM_005256219.3 | 4 | 1883 | XP_005256276.1 | |
XM_011523424.2 | 5 | 1842 | XP_011521726.1 | |
XM_017023843.1 | 4 | 2007 | XP_016879332.1 | |
AF064599.1 | 4 | 1211 | AAC18593.1 | |
BC012798.2 | 4 | 1256 | AAH12798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)