Assay Details
Target Gene Details
Entrez Gene ID: | 54880 |
Gene Name: | BCL6 corepressor |
Gene Aliases: |
ANOP2, MAA2, MCOPS2 |
Location: |
Chr.X:40051246-40177390 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCOR | NM_001123383.1 | 15 | 5526 | NP_001116855.1 |
NM_001123384.1 | 14 | 5472 | NP_001116856.1 | |
NM_001123385.1 | 15 | 5557 | NP_001116857.1 | |
NM_017745.5 | 15 | 5455 | NP_060215.4 | |
XM_005272616.1 | 15 | 5689 | XP_005272673.1 | |
XM_005272618.3 | 15 | 5355 | XP_005272675.1 | |
XM_005272619.3 | 14 | 5961 | XP_005272676.1 | |
XM_005272620.3 | 14 | 5859 | XP_005272677.1 | |
XM_006724536.3 | 15 | 6791 | XP_006724599.1 | |
XM_011543929.2 | 15 | 5465 | XP_011542231.1 | |
XM_011543930.1 | 15 | 5478 | XP_011542232.1 | |
XM_011543931.2 | 15 | 5588 | XP_011542233.1 | |
XM_017029615.1 | 15 | 6689 | XP_016885104.1 | |
XM_017029616.1 | 15 | 6063 | XP_016885105.1 | |
AF317391.1 | 15 | 5414 | AAG41429.1 | |
AK000292.1 | 7 | 1332 | BAA91061.1 | |
AK074286.1 | 11 | 4463 | BAB85037.1 | |
AK095360.1 | 10 | 2166 | ||
AK225581.1 | 7 | 1285 | ||
AY316592.1 | 15 | 5516 | AAR08265.1 | |
BC009675.2 | 6 | 1083 | AAH09675.2 | |
BC063536.1 | 4 | 639 | AAH63536.1 | |
BC114220.1 | 14 | 5472 | AAI14221.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |