Assay Details
Target Gene Details
Entrez Gene ID: | 55695 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 5 |
Gene Aliases: |
NOL1, NOL1R, NSUN5A, WBSCR20, WBSCR20A, p120, p120(NOL1) |
Location: |
Chr.7:73302516-73308867 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN5 | NM_001168347.2 | NP_001161819.1 | ||
NM_001168348.2 | NP_001161820.1 | |||
NM_018044.4 | NP_060514.1 | |||
NM_148956.3 | NP_683759.1 | |||
XM_005277604.4 | XP_005277661.1 | |||
AF412028.1 | AAM62310.1 | |||
AF420249.1 | AAL16067.1 | |||
AK001129.1 | BAA91515.1 | |||
AK098619.1 | ||||
AK125667.1 | 2 | 254 | ||
AK126375.1 | ||||
AK298221.1 | ||||
AK315361.1 | ||||
BC008084.2 | AAH08084.1 | |||
CB154882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759537 | Chr.7:72446485 - 73461568 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1161563 | Chr.7:72932296 - 73309388 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv6466n100 | Chr.7:73245562 - 73420814 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv831027 | Chr.7:73173922 - 73442066 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv970531 | Chr.7:73291991 - 73324627 on Build GRCh38 | Duplication |
![]() ![]() |
esv34349 | Chr.7:72578056 - 73341521 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)