Assay Details
Target Gene Details
Entrez Gene ID: | 5118 |
Gene Name: | procollagen C-endopeptidase enhancer |
Gene Aliases: |
PCPE, PCPE-1, PCPE1 |
Location: |
Chr.7:100602259-100608175 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCOLCE | NM_002593.3 | NP_002584.2 | ||
AB008549.1 | BAA23281.1 | |||
AK298364.1 | ||||
AK313748.1 | ||||
BC000574.2 | AAH00574.1 | |||
BC033205.1 | AAH33205.1 | |||
BI837971.1 | ||||
L33799.1 | AAA61949.1 |
Target Gene Details
Entrez Gene ID: | 100129845 |
Gene Name: | PCOLCE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:100589401-100604038 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCOLCE-AS1 | NR_038910.1 | 2 | 806 | |
AK097705.1 | 2 | 806 | ||
AK298216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2659884 | Chr.7:100600622 - 100605969 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)