Assay Details
Target Gene Details
Entrez Gene ID: | 2997 |
Gene Name: | glycogen synthase 1 |
Gene Aliases: |
GSY, GYS |
Location: |
Chr.19:48968125-48993353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GYS1 | NM_001161587.1 | NP_001155059.1 | ||
NM_002103.4 | NP_002094.2 | |||
NR_027763.1 | ||||
AK223579.1 | BAD97299.1 | |||
AK291672.1 | ||||
AK294349.1 | ||||
AK300369.1 | ||||
AK302738.1 | ||||
AK303712.1 | ||||
BC002617.2 | AAH02617.1 | |||
BC003182.1 | AAH03182.1 | |||
BC007688.2 | AAH07688.1 | |||
J04501.1 | AAA88046.1 | |||
U32573.1 | AAB60385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | NUP62 LIN7B LIG1 SNAR-C4 FCGRT SNAR-G2 PRR12 PTOV1-AS1 FAM83E SNORD32A MIR4750 SNAR-A2 SPACA4 MAMSTR SLC17A7 SNORD35A NTN5 FTL C19orf68 SIGLEC11 VRK3 MIR6798 BCAT2 MIR4324 SNAR-A14 MIR5088 CYTH2 CGB5 LOC101059948 TSKS RPL18 GYS1 SCAF1 SNORD34 CGB1 NTF4 LOC105372430 KCNA7 PLEKHA4 PRMT1 TULP2 CGB7 MIR4749 MIR4751 SNORD33 PLA2G4C SNAR-A3 C19orf73 HSD17B14 CARD8-AS1 RUVBL2 SNAR-G1 ELSPBP1 SEC1P BAX SLC6A16 RASIP1 LOC105447645 IL4I1 PTOV1 SNRNP70 RPL13A CGB3 ALDH16A1 SNAR-A6 PTH2 GRIN2D AP2A1 SNAR-A7 CA11 SNAR-D ATF5 MIR150 DBP CCDC114 MED25 FUZ KDELR1 EMP3 SNAR-C1 SNAR-C5 SNORD35B GRWD1 GFY CPT1C SPHK2 TEAD2 SULT2B1 MIR6800 ADM5 FUT2 MIR6799 SNAR-A5 CARD8 PLA2G4C-AS1 NUCB1 SNAR-A9 SYNGR4 ZNF473 SNAR-C2 PNKP FLJ26850 RPS11 LOC101928295 PIH1D1 NUCB1-AS1 TMEM143 LMTK3 SNAR-C3 CGB8 DKKL1 RCN3 PRRG2 SNAR-A10 ZNF114 CGB2 FLT3LG DHDH SNAR-B1 SNAR-A4 TRPM4 NOSIP LOC100287477 FUT1 PPFIA3 SNAR-A12 PPP1R15A BSPH1 SIGLEC16 HRC IZUMO1 BCL2L12 IRF3 CCDC155 SNAR-B2 RRAS AKT1S1 LHB SNAR-A11 CABP5 SNAR-A13 PTOV1-AS2 FGF21 SNAR-A8 TBC1D17 CD37 SNAR-A1 KCNJ14 |
dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain | RUVBL2 FTL LHB BAX LOC101059948 MIR6798 GYS1 |
nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss | SNRNP70 CGB2 CGB3 CGB7 LIN7B LHB DHDH CGB5 LOC101059948 SNAR-G2 GYS1 NUCB1-AS1 C19orf73 RUVBL2 SNAR-G1 PPFIA3 FTL CGB8 BAX MIR6798 CGB1 NTF4 NUCB1 KCNA7 |
nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion | SNRNP70 CGB2 CGB3 CGB7 LHB CGB5 LOC101059948 SNAR-G2 GYS1 RUVBL2 SNAR-G1 FTL CGB8 BAX MIR6798 CGB1 NTF4 KCNA7 |
nsv833856 | Chr.19:48847866 - 48986483 on Build GRCh38 | Loss | PLEKHA4 TULP2 FTL PPP1R15A BAX DHDH GYS1 NUCB1 NUCB1-AS1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |