Assay Details
Target Gene Details
Entrez Gene ID: | 84812 |
Gene Name: | phospholipase C delta 4 |
Gene Aliases: |
- |
Location: |
Chr.2:218606873-218637186 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCD4 | NM_032726.3 | NP_116115.1 | ||
XM_005246913.2 | XP_005246970.1 | |||
XM_011512012.1 | XP_011510314.1 | |||
XM_017005115.1 | XP_016860604.1 | |||
XM_017005116.1 | XP_016860605.1 | |||
XM_017005117.1 | XP_016860606.1 | |||
XM_017005118.1 | XP_016860607.1 | |||
AK023083.1 | ||||
AK057186.1 | ||||
AK223203.1 | BAD96923.1 | |||
AK297808.1 | ||||
AK299458.1 | ||||
AY512961.1 | AAS82574.1 | |||
BC006355.2 | AAH06355.1 | |||
BP361394.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |