Assay Details
Target Gene Details
Entrez Gene ID: | 84435 |
Gene Name: | adhesion G protein-coupled receptor A1 |
Gene Aliases: |
GPR123 |
Location: |
Chr.10:133070929-133131675 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRA1 | XM_005252695.1 | 3 | 555 | XP_005252752.1 |
AY181241.1 | 3 | 555 | AAO27353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759793 | Chr.10:133030918 - 133619181 on Build GRCh38 | Gain+Loss | MIR3944 ZNF511 TUBGCP2 ADGRA1-AS1 ADGRA1 MIR202HG PRAP1 CYP2E1 SPRNP1 CALY KNDC1 SCART1 ECHS1 PAOX ADAM8 SPRN FUOM UTF1 MIR202 SYCE1 MTG1 VENTX |
dgv998n100 | Chr.10:132998894 - 133170978 on Build GRCh38 | Gain | ADGRA1-AS1 ADGRA1 KNDC1 |
esv3625022 | Chr.10:132998655 - 133120915 on Build GRCh38 | Gain | ADGRA1-AS1 ADGRA1 |
nsv467498 | Chr.10:132900981 - 133134139 on Build GRCh38 | Gain | ADGRA1-AS1 LINC01166 ADGRA1 CFAP46 LINC01168 LINC01167 |
esv3625015 | Chr.10:132889012 - 133114687 on Build GRCh38 | Gain | ADGRA1-AS1 LINC01166 ADGRA1 CFAP46 LINC01168 LINC01167 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | ADGRA1-AS1 PWWP2B ADGRA1 TCERG1L-AS1 LRRC27 MIR378C LOC107984282 STK32C LINC01166 LINC01164 CFAP46 JAKMIP3 DPYSL4 C10orf91 TCERG1L LINC01168 BNIP3 LOC105378568 INPP5A PPP2R2D LINC01167 NKX6-2 |
nsv528260 | Chr.10:132859348 - 133620799 on Build GRCh38 | Loss | ZNF511 TUBGCP2 MIR202HG CYP2E1 CALY KNDC1 SPRN CFAP46 UTF1 LINC01167 MIR3944 ADGRA1-AS1 ADGRA1 PRAP1 SPRNP1 SCART1 ECHS1 PAOX ADAM8 LINC01166 FUOM MIR202 LINC01168 SYCE1 MTG1 VENTX |
nsv952703 | Chr.10:133037897 - 133143896 on Build GRCh38 | Deletion | ADGRA1-AS1 ADGRA1 |
nsv470979 | Chr.10:133033478 - 133118280 on Build GRCh38 | Loss | ADGRA1-AS1 ADGRA1 |
nsv552526 | Chr.10:132699017 - 133103793 on Build GRCh38 | Gain | ADGRA1-AS1 LINC01166 ADGRA1 CFAP46 LINC01168 INPP5A LINC01167 NKX6-2 |
nsv552539 | Chr.10:133061918 - 133103793 on Build GRCh38 | Loss | ADGRA1-AS1 ADGRA1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |