Assay Details
Target Gene Details
Entrez Gene ID: | 5331 |
Gene Name: | phospholipase C beta 3 |
Gene Aliases: |
- |
Location: |
Chr.11:64251523-64269452 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCB3 | NM_000932.2 | NP_000923.1 | ||
NM_001184883.1 | NP_001171812.1 | |||
NM_001316314.1 | NP_001303243.1 | |||
XM_011545101.2 | XP_011543403.1 | |||
XM_017017925.1 | XP_016873414.1 | |||
BC032659.1 | AAH32659.1 | |||
BC142681.1 | ||||
Z16411.1 | CAA78903.1 | |||
Z26649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | NUDT22 TRPT1 FERMT3 PLCB3 GPR137 VEGFB BAD PPP1R14B STIP1 FKBP2 LOC105369340 DNAJC4 MACROD1 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | KCNK4 MIR7155 NUDT22 TRPT1 FERMT3 PLCB3 PRDX5 VEGFB BAD COX8A TRMT112 OTUB1 STIP1 MACROD1 ESRRA CCDC88B GPR137 FLRT1 PPP1R14B TEX40 NAA40 FKBP2 LOC105369340 DNAJC4 KCNK4-TEX40 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | ESRRA KCNK4 NUDT22 TRPT1 FERMT3 PLCB3 GPR137 PRDX5 VEGFB BAD PPP1R14B TRMT112 TEX40 STIP1 FKBP2 LOC105369340 DNAJC4 KCNK4-TEX40 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | PPP1R14B NUDT22 TRPT1 FERMT3 STIP1 PLCB3 FKBP2 LOC105369340 DNAJC4 VEGFB |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | ESRRA KCNK4 CCDC88B MIR7155 NUDT22 TRPT1 FERMT3 PLCB3 RPS6KA4 MIR1237 GPR137 PRDX5 VEGFB BAD PPP1R14B TRMT112 TEX40 FKBP2 LOC105369340 DNAJC4 KCNK4-TEX40 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | NUDT22 TRPT1 FERMT3 PLCB3 GPR137 VEGFB BAD PPP1R14B STIP1 FKBP2 LOC105369340 DNAJC4 MACROD1 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | ESRRA KCNK4 NUDT22 TRPT1 FERMT3 PLCB3 GPR137 PRDX5 VEGFB BAD FLRT1 PPP1R14B TRMT112 TEX40 STIP1 FKBP2 LOC105369340 DNAJC4 KCNK4-TEX40 MACROD1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |