Assay Details
Target Gene Details
Entrez Gene ID: | 1524 |
Gene Name: | C-X3-C motif chemokine receptor 1 |
Gene Aliases: |
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28 |
Location: |
Chr.3:39263494-39281735 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CX3CR1 | NM_001171171.1 | 2 | 332 | NP_001164642.1 |
NM_001171172.1 | 2 | 292 | NP_001164643.1 | |
NM_001171174.1 | 2 | 228 | NP_001164645.1 | |
NM_001337.3 | 2 | 184 | NP_001328.1 | |
AK225764.1 | 2 | 184 | ||
AK307911.1 | ||||
AK312373.1 | 2 | 183 | ||
BC028078.1 | 2 | 168 | AAH28078.1 | |
DA396634.1 | 2 | 174 | ||
DA406242.1 | 2 | 332 | ||
DB274136.1 | 2 | 292 | ||
U20350.1 | 2 | 179 | AAA91783.1 | |
U28934.1 | 2 | 137 | AAA87032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3772 | Chr.3:39240511 - 39273967 on Build GRCh38 | Insertion | CX3CR1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |