Assay Details
Target Gene Details
Entrez Gene ID: | 339501 |
Gene Name: | protease, serine 38 |
Gene Aliases: |
MPN2 |
Location: |
Chr.1:227815708-227846470 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS38 | NM_183062.2 | 5 | 1124 | NP_898885.1 |
AA398170.1 | 1 | 121 | ||
BC130400.1 | 5 | 1196 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549273 | Chr.1:227808265 - 227863533 on Build GRCh38 | Loss | PRSS38 |
nsv826908 | Chr.1:227707685 - 228503399 on Build GRCh38 | Gain | JMJD4 RNF187 LOC100506571 LOC101060022 GJC2 HIST3H2A LOC107985355 HIST3H2BB TRIM17 GUK1 OBSCN IBA57 C1orf145 MRPL55 MIR6742 MIR3620 MIR5008 SNAP47 LOC101927401 IBA57-AS1 ARF1 WNT9A PRSS38 C1orf35 TRIM11 WNT3A LOC105373289 MIR4666A HIST3H3 |
esv32853 | Chr.1:227692415 - 228670462 on Build GRCh38 | Gain | JMJD4 RNA5S13 RNA5S11 RNA5S8 BTNL10 HIST3H2A RNA5S10 TRIM17 GUK1 RNA5S6 IBA57 C1orf145 RNA5S5 RNA5S2 MIR3620 SNAP47 ARF1 RNA5S3 C1orf35 RNA5S12 RNA5S1 WNT3A MIR4666A RNA5S4 RNA5S15 RNF187 LOC100506571 LOC101060022 RNA5S17 RNA5S14 GJC2 RNA5S7 LOC107985355 HIST3H2BB OBSCN MRPL55 RNA5S16 RHOU MIR6742 RNA5S9 MIR5008 LOC101927401 IBA57-AS1 WNT9A PRSS38 TRIM11 LOC105373132 LOC105373289 DUSP5P1 HIST3H3 |
nsv468249 | Chr.1:227828669 - 227857807 on Build GRCh38 | Loss | PRSS38 |
nsv998933 | Chr.1:227805227 - 227871595 on Build GRCh38 | Loss | PRSS38 |
nsv549272 | Chr.1:227748061 - 227858047 on Build GRCh38 | Loss | LOC105373289 SNAP47 PRSS38 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |