Assay Details
Target Gene Details
Entrez Gene ID: | 87 |
Gene Name: | actinin alpha 1 |
Gene Aliases: |
BDPLT15 |
Location: |
Chr.14:68874123-68979366 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACTN1 | NM_001102.3 | 21 | 3044 | NP_001093.1 |
NM_001130004.1 | 22 | 3110 | NP_001123476.1 | |
NM_001130005.1 | 21 | 3029 | NP_001123477.1 | |
XM_011537265.2 | 21 | 2860 | XP_011535567.1 | |
XM_011537266.2 | 21 | 2845 | XP_011535568.1 | |
XM_011537267.2 | 21 | 2836 | XP_011535569.1 | |
XM_011537268.2 | 21 | 2821 | XP_011535570.1 | |
XM_017021720.1 | 21 | 3391 | XP_016877209.1 | |
XM_017021721.1 | 21 | 3367 | XP_016877210.1 | |
XM_017021722.1 | 21 | 3536 | XP_016877211.1 | |
XM_017021723.1 | 21 | 3512 | XP_016877212.1 | |
XM_017021725.1 | 21 | 3710 | XP_016877214.1 | |
XM_017021726.1 | 21 | 3686 | XP_016877215.1 | |
XM_017021727.1 | 21 | 2990 | XP_016877216.1 | |
XM_017021728.1 | 21 | 3140 | XP_016877217.1 | |
AK098203.1 | 14 | 1762 | ||
AK124378.1 | 2 | 2155 | ||
AK225782.1 | 21 | 2926 | ||
AK294317.1 | 15 | 2232 | ||
AK295099.1 | 21 | 2925 | ||
AK298496.1 | ||||
BC003576.1 | 21 | 2976 | AAH03576.1 | |
BC015766.1 | 21 | 2851 | AAH15766.1 | |
BC127124.1 | ||||
BT007207.1 | AAP35871.1 | |||
BX641076.1 | 10 | 4183 | ||
DQ496098.1 | 22 | 2983 | ||
EU716325.1 | ||||
FJ410030.1 | ||||
M95178.1 | AAA51582.1 | |||
X15804.1 | 21 | 2932 | CAA33803.1 | |
X55187.1 | 12 | 1846 | CAA38970.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |