Assay Details
Target Gene Details
Entrez Gene ID: | 56124 |
Gene Name: | protocadherin beta 12 |
Gene Aliases: |
PCDH-BETA12 |
Location: |
Chr.5:141208719-141212126 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCDHB12 | NM_018932.3 | 1 | 121 | NP_061755.1 |
AK293336.1 | 1 | 143 | ||
AK316160.1 | 1 | 121 | ||
BC045637.1 | 1 | 121 | AAH45637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv599919 | Chr.5:141182193 - 141212836 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3606974 | Chr.5:141203379 - 141210938 on Build GRCh38 | Gain |
![]() |
nsv969261 | Chr.5:141199392 - 141210803 on Build GRCh38 | Duplication |
![]() ![]() |
nsv823266 | Chr.5:141156966 - 141222863 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2730846 | Chr.5:141193679 - 141210136 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv1024594 | Chr.5:141167591 - 141243953 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471634 | Chr.5:141106394 - 141258375 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428127 | Chr.5:141016768 - 141319173 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2664767 | Chr.5:141177668 - 141213756 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv958e201 | Chr.5:141174165 - 141215536 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv599908 | Chr.5:141139584 - 141250122 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)