Assay Details
Target Gene Details
Entrez Gene ID: | 201501 |
Gene Name: | zinc finger and BTB domain containing 7C |
Gene Aliases: |
APM-1, APM1, ZBTB36, ZNF857C |
Location: |
Chr.18:48026886-48412639 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB7C | NM_001039360.2 | 2 | 945 | NP_001034449.1 |
NM_001318841.1 | 4 | 1126 | NP_001305770.1 | |
XM_005258229.4 | 3 | 870 | XP_005258286.1 | |
XM_011525861.2 | 6 | 1350 | XP_011524163.1 | |
XM_011525863.2 | 5 | 1467 | XP_011524165.1 | |
XM_011525864.2 | 5 | 1222 | XP_011524166.1 | |
XM_011525865.2 | 5 | 1463 | XP_011524167.1 | |
XM_011525866.2 | 4 | 1208 | XP_011524168.1 | |
XM_011525867.2 | 4 | 997 | XP_011524169.1 | |
XM_011525869.2 | 5 | 1472 | XP_011524171.1 | |
XM_011525870.2 | 4 | 1168 | XP_011524172.1 | |
XM_011525871.2 | 2 | 1683 | XP_011524173.1 | |
XM_017025605.1 | 6 | 3145 | XP_016881094.1 | |
XM_017025606.1 | 6 | 1599 | XP_016881095.1 | |
XM_017025607.1 | 6 | 1354 | XP_016881096.1 | |
XM_017025608.1 | 5 | 3022 | XP_016881097.1 | |
XM_017025609.1 | 4 | 1339 | XP_016881098.1 | |
AK296709.1 | 2 | 945 | ||
AM988836.1 | 2 | 933 | ||
AM988837.1 | 2 | 1078 | ||
AM988838.1 | 3 | 1128 | ||
AM988839.1 | 4 | 1126 | ||
EF120357.1 | 2 | 997 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |