Assay Details
Target Gene Details
Entrez Gene ID: | 26261 |
Gene Name: | F-box protein 24 |
Gene Aliases: |
FBX24 |
Location: |
Chr.7:100582490-100601117 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXO24 | NM_001163499.1 | NP_001156971.1 | ||
NM_012172.4 | NP_036304.2 | |||
NM_033506.2 | NP_277041.1 | |||
XM_005250259.3 | XP_005250316.1 | |||
XM_011516022.1 | XP_011514324.1 | |||
XM_011516023.2 | XP_011514325.1 | |||
XM_017011961.1 | XP_016867450.1 | |||
AK301864.1 | ||||
AL136811.1 | CAB66745.1 | |||
BC067293.1 | AAH67293.1 |
Target Gene Details
Entrez Gene ID: | 100129845 |
Gene Name: | PCOLCE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:100589401-100604038 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCOLCE-AS1 | NR_038910.1 | |||
AK097705.1 | ||||
AK298216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2659884 | Chr.7:100600622 - 100605969 on Build GRCh38 | Deletion | PCOLCE-AS1 PCOLCE FBXO24 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | ZSCAN21 PILRA PCOLCE ZCWPW1 FBXO24 AP1S1 SLC12A9 SERPINE1 GATS C7orf61 SPDYE3 UFSP1 PPP1R35 GPC2 MUC17 MUC12 FIS1 POP7 CNPY4 NYAP1 AZGP1P1 STAG3 LOC102724094 MOSPD3 AZGP1 ACTL6B CLDN15 ZNHIT1 TFR2 GNB2 ZNF3 TRIP6 MIR4653 ZKSCAN1 EPHB4 VGF STAG3L5P TSC22D4 MEPCE MIR6840 MBLAC1 MCM7 AGFG2 EPO ZAN PLOD3 MIR106B MIR93 GIGYF1 TAF6 PCOLCE-AS1 LOC105375429 LAMTOR4 PILRB GAL3ST4 TRIM56 ZASP PVRIG AP4M1 SAP25 SRRT MIR25 PMS2P1 LRCH4 ACHE MUC3A NAT16 MOGAT3 PVRIG2P STAG3L5P-PVRIG2P-PILRB MIR6875 MIR4658 C7orf43 COPS6 |
nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion | PCOLCE SAP25 MOSPD3 FBXO24 LRCH4 AGFG2 GIGYF1 PCOLCE-AS1 ACTL6B TFR2 GNB2 LOC105375429 ZASP |
nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss | POP7 PCOLCE SAP25 MOSPD3 FBXO24 LRCH4 AGFG2 EPO ZAN GIGYF1 PCOLCE-AS1 ACTL6B TFR2 GNB2 LOC105375429 ZASP |
nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion | PCOLCE-AS1 PCOLCE SAP25 MOSPD3 FBXO24 LRCH4 ZASP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |