Assay Details
Target Gene Details
Entrez Gene ID: | 11218 |
Gene Name: | DEAD-box helicase 20 |
Gene Aliases: |
DP103, GEMIN3 |
Location: |
Chr.1:111755568-111767577 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX20 | NM_007204.4 | 5 | 1092 | NP_009135.4 |
AF106019.1 | 5 | 745 | AAD42744.1 | |
AF155803.1 | 5 | 740 | AAK56799.1 | |
AF171063.1 | 5 | 1028 | AAF14544.1 | |
AK001506.1 | 5 | 754 | BAA91727.1 | |
AK301697.1 | 4 | 593 | ||
AK314312.1 | 5 | 737 | ||
AL133598.1 | 2 | 247 | CAB63734.2 | |
BC011556.1 | 5 | 745 | AAH11556.1 | |
BC031062.1 | 5 | 754 | AAH31062.1 | |
BC034953.2 | 5 | 1063 | AAH34953.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |