Assay Details
Target Gene Details
Entrez Gene ID: | 4015 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
- |
Location: |
Chr.5:122063195-122078501 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOX | NM_001178102.2 | NP_001171573.1 | ||
NM_001317073.1 | NP_001304002.1 | |||
NM_002317.6 | NP_002308.2 | |||
AF039291.1 | AAD02130.1 | |||
AK297709.1 | ||||
AK298781.1 | ||||
AK307559.1 | ||||
AK307732.1 | ||||
AK311354.1 | ||||
AK312269.1 | ||||
AK315873.1 | ||||
AK316309.1 | ||||
BC074820.2 | AAH74820.1 | |||
BC074872.2 | AAH74872.1 | |||
BC089436.1 | AAH89436.1 | |||
CN265862.1 | ||||
M94054.1 | AAA59525.1 | |||
S45875.1 | AAB23549.1 | |||
S78694.1 | AAB21243.1 |
Target Gene Details
Entrez Gene ID: | 153443 |
Gene Name: | serum response factor binding protein 1 |
Gene Aliases: |
BUD22, P49, Rlb1, STRAP, p49/STRAP |
Location: |
Chr.5:121961917-122075624 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRFBP1 | XM_017009111.1 | XP_016864600.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |