Assay Details
Target Gene Details
Entrez Gene ID: | 56834 |
Gene Name: | G protein-coupled receptor 137 |
Gene Aliases: |
C11orf4, GPR137A, TM7SF1L1 |
Location: |
Chr.11:64270337-64289500 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv357 | Chr.11:64284335 - 64308789 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)