Assay Details
Target Gene Details
Entrez Gene ID: | 6531 |
Gene Name: | solute carrier family 6 member 3 |
Gene Aliases: |
DAT, DAT1, PKDYS |
Location: |
Chr.5:1392790-1445428 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC6A3 | NM_001044.4 | 15 | 2644 | NP_001035.1 |
AK310589.1 | 3 | 899 | ||
BC132977.1 | 14 | 2569 | ||
BC133003.1 | 14 | 2569 | ||
EF174603.1 | 15 | 2646 | ||
M95167.1 | 15 | 2646 | AAC41720.1 | |
S46955.1 | 15 | 2619 | AAA11754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv9535n54 | Chr.5:1370717 - 1394962 on Build GRCh38 | Loss | LINC01511 SLC6A3 |
nsv950628 | Chr.5:1380586 - 1414985 on Build GRCh38 | Deletion | SLC6A3 |
esv3603807 | Chr.5:1384232 - 1416519 on Build GRCh38 | Loss | SLC6A3 |
nsv830187 | Chr.5:1317991 - 1509711 on Build GRCh38 | Loss | LPCAT1 CLPTM1L LINC01511 SLC6A3 |
nsv596853 | Chr.5:1308437 - 1394962 on Build GRCh38 | Gain | MIR4457 CLPTM1L LINC01511 SLC6A3 |
nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain | SLC12A7 NKD2 CLPTM1L TRIP13 LOC100996325 TERT SLC6A3 MIR4456 MIR4457 SLC9A3 MIR4635 LINC01511 BRD9 SLC6A18 LPCAT1 EXOC3 ZDHHC11 PP7080 CTD-3080P12.3 TPPP CEP72 LOC101929895 ZDHHC11B LOC100506688 SLC6A19 LOC100288152 |
nsv509045 | Chr.5:1282173 - 1455603 on Build GRCh38 | Insertion | MIR4457 CLPTM1L LINC01511 TERT SLC6A3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |