Assay Details
Target Gene Details
Entrez Gene ID: | 337880 |
Gene Name: | keratin associated protein 11-1 |
Gene Aliases: |
HACL-1, HACL1, KAP11.1 |
Location: |
Chr.21:30880644-30881555 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KRTAP11-1 | NM_175858.2 | 1 | 514 | NP_787054.1 |
AB096963.1 | BAE46378.1 | |||
AJ457065.1 | 1 | 514 | CAD29721.1 | |
BC130555.1 | 1 | 507 | ||
BC130557.1 | 1 | 507 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834078 | Chr.21:30847656 - 31014506 on Build GRCh38 | Gain |
![]() |
nsv834077 | Chr.21:30758503 - 30921578 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv518454 | Chr.21:30842336 - 30927564 on Build GRCh38 | Loss |
![]() |
nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3646842 | Chr.21:30849249 - 30930214 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)