Assay Details
Target Gene Details
Entrez Gene ID: | 54617 |
Gene Name: | INO80 complex subunit |
Gene Aliases: |
INO80A, INOC1, hINO80 |
Location: |
Chr.15:40978880-41117081 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INO80 | NM_017553.2 | 36 | 5174 | NP_060023.1 |
NR_104038.1 | 35 | 5046 | ||
XM_011521686.2 | 22 | 2985 | XP_011519988.1 | |
AB033085.1 | 36 | 5070 | ||
AK002176.1 | 8 | 1338 | BAA92122.1 | |
AK024777.1 | BAB14997.1 | |||
AL137280.1 | 5 | 954 | CAB70675.1 | |
BC146785.1 | 36 | 5082 | ||
BX350023.2 | 1 | 161 | ||
CB155560.1 | 1 | 344 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053462 | Chr.15:40977869 - 41103896 on Build GRCh38 | Gain | INO80 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |