Assay Details
Target Gene Details
Entrez Gene ID: | 529 |
Gene Name: | ATPase H+ transporting V1 subunit E1 |
Gene Aliases: |
ATP6E, ATP6E2, ATP6V1E, P31, Vma4 |
Location: |
Chr.22:17592136-17628822 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V1E1 | NM_001039366.1 | NP_001034455.1 | ||
NM_001039367.1 | NP_001034456.1 | |||
NM_001696.3 | NP_001687.1 | |||
AK294623.1 | ||||
AK312394.1 | ||||
AK315941.1 | ||||
BC004443.1 | AAH04443.1 | |||
BE735148.1 | ||||
BI546032.1 | ||||
BI597419.1 | ||||
BQ888820.1 | ||||
BT007128.1 | AAP35792.1 | |||
CK005516.1 | ||||
CR456385.1 | CAG30271.1 | |||
X71491.1 | CAA50592.1 | |||
X76228.1 | CAA53814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | LOC101929372 BCL2L13 ATP6V1E1 SLC25A18 CECR2 |
dgv4464n100 | Chr.22:17570790 - 17611340 on Build GRCh38 | Gain | LOC101929372 ATP6V1E1 SLC25A18 |
nsv1065055 | Chr.22:17559085 - 17632626 on Build GRCh38 | Gain | LOC101929372 BCL2L13 ATP6V1E1 SLC25A18 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | MICAL3 LOC101929372 BCL2L13 ATP6V1E1 SLC25A18 MIR3198-1 LINC00528 BID CECR2 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | FLJ41941 MICAL3 MIR648 LOC101929372 BCL2L13 ATP6V1E1 SLC25A18 MIR3198-1 LINC00528 BID |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | LOC101929372 BCL2L13 ATP6V1E1 SLC25A18 |
nsv3553 | Chr.22:17553601 - 17608096 on Build GRCh38 | Deletion | LOC101929372 ATP6V1E1 SLC25A18 CECR2 |
nsv510802 | Chr.22:17550100 - 17604891 on Build GRCh38 | Deletion | LOC101929372 ATP6V1E1 SLC25A18 CECR2 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC100996401 MICAL3 GGT3P LOC100996415 BCL2L13 PI4KAP1 LOC729461 CECR2 MIR648 LOC101929372 ATP6V1E1 SLC25A18 TUBA8 PEX26 LOC102725072 LOC642643 LINC00528 BID LOC100996432 FLJ41941 TMEM191B USP18 RIMBP3 MIR3198-1 FAM230A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |