Assay Details
Target Gene Details
Entrez Gene ID: | 8863 |
Gene Name: | period circadian clock 3 |
Gene Aliases: |
FASPS3, GIG13 |
Location: |
Chr.1:7784285-7845181 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PER3 | NM_001289861.1 | 2 | 145 | NP_001276790.1 |
NM_001289862.1 | 2 | 145 | NP_001276791.1 | |
NM_001289863.1 | 2 | 145 | NP_001276792.1 | |
NM_001289864.1 | 2 | 145 | NP_001276793.1 | |
NM_016831.2 | 2 | 145 | NP_058515.1 | |
XM_005263524.3 | XP_005263581.1 | |||
XM_011542385.2 | XP_011540687.1 | |||
XM_011542390.2 | XP_011540692.1 | |||
XM_017002723.1 | XP_016858212.1 | |||
XM_017002724.1 | XP_016858213.1 | |||
XM_017002725.1 | XP_016858214.1 | |||
XM_017002726.1 | XP_016858215.1 | |||
XM_017002727.1 | 2 | 197 | XP_016858216.1 | |
XM_017002728.1 | 2 | 197 | XP_016858217.1 | |
XM_017002730.1 | 2 | 197 | XP_016858219.1 | |
XM_017002731.1 | 2 | 197 | XP_016858220.1 | |
XM_017002732.1 | 1 | 246 | XP_016858221.1 | |
XM_017002733.1 | 2 | 197 | XP_016858222.1 | |
XM_017002734.1 | 2 | 197 | XP_016858223.1 | |
XM_017002735.1 | 2 | 197 | XP_016858224.1 | |
XM_017002736.1 | 2 | 197 | XP_016858225.1 | |
AB047686.1 | 1 | 105 | ||
AK307760.1 | ||||
AK308303.1 | 2 | 194 | ||
AL157954.1 | 1 | 56 | CAB76084.1 | |
BC026102.1 | 2 | 167 | AAH26102.1 | |
BC146781.1 | 1 | 105 | ||
HY151851.1 | 2 | 145 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |