Assay Details
Target Gene Details
Entrez Gene ID: | 26019 |
Gene Name: | UPF2 regulator of nonsense transcripts homolog (yeast) |
Gene Aliases: |
HUPF2, RENT2, smg-3 |
Location: |
Chr.10:11920022-12043170 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UPF2 | NM_015542.3 | 22 | 4234 | NP_056357.1 |
NM_080599.2 | 22 | 4404 | NP_542166.1 | |
XM_017016034.1 | 21 | 3890 | XP_016871523.1 | |
AB037829.1 | 21 | 4603 | ||
AF301013.1 | 22 | 4227 | AAG33225.1 | |
AF318574.1 | 22 | 4204 | AAG60689.1 | |
AK000764.1 | 7 | 1065 | BAA91369.1 | |
AK074375.1 | 7 | 999 | BAB85064.1 | |
AK096191.1 | 9 | 1881 | BAC04721.1 | |
AK124930.1 | 1 | 1292 | ||
AK295209.1 | ||||
AL080198.1 | 16 | 2404 | CAB45771.1 | |
BC114964.1 | 22 | 4198 | ||
BC115737.1 | 22 | 4198 | ||
BX647325.1 | 1 | 1217 |
More Information
Additional Information:
For this assay, SNP(s) [rs74119905] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |