Assay Details
Target Gene Details
Entrez Gene ID: | 160364 |
Gene Name: | C-type lectin domain family 12 member A |
Gene Aliases: |
CD371, CLL-1, CLL1, DCAL-2, MICL |
Location: |
Chr.12:9951316-9995209 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLEC12A | NM_001207010.1 | NP_001193939.1 | ||
NM_001300730.1 | NP_001287659.1 | |||
NM_138337.5 | NP_612210.4 | |||
NM_201623.3 | NP_963917.2 | |||
XM_005253324.2 | XP_005253381.1 | |||
XM_006719033.3 | XP_006719096.1 | |||
XM_006719035.3 | XP_006719098.1 | |||
XM_006719036.3 | XP_006719099.1 | |||
XM_011520570.1 | XP_011518872.1 | |||
XM_011520571.2 | XP_011518873.2 | |||
XM_011520573.2 | XP_011518875.1 | |||
AF247788.1 | AAL95693.1 | |||
AK314001.1 | ||||
AY426759.1 | AAR84594.1 | |||
AY498550.1 | AAS00605.1 | |||
AY498551.1 | AAS00606.1 | |||
AY498552.1 | AAS00607.1 | |||
AY547296.1 | AAT11783.1 | |||
BC063424.1 | AAH63424.1 | |||
BC126289.1 | ||||
BC126291.1 | ||||
CD642076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1038226 | Chr.12:9968689 - 9990992 on Build GRCh38 | Gain | CLEC12A CLEC1B |
nsv525593 | Chr.12:9968748 - 10004047 on Build GRCh38 | Loss | CLEC12A CLEC1B |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |