Assay Details
Target Gene Details
Entrez Gene ID: | 2532 |
Gene Name: | atypical chemokine receptor 1 (Duffy blood group) |
Gene Aliases: |
CCBP1, CD234, DARC, Dfy, FY, GPD, GpFy, WBCQ1 |
Location: |
Chr.1:159204013-159206500 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACKR1 | NM_001122951.2 | 1 | 754 | NP_001116423.1 |
NM_002036.3 | 2 | 1520 | NP_002027.2 | |
AF030521.1 | 1 | 579 | AAD20435.1 | |
AK291593.1 | 2 | 1520 | ||
AK309521.1 | 1 | 1023 | ||
AY167991.1 | 1 | 573 | AAO11657.1 | |
BC017817.1 | 2 | 725 | AAH17817.1 | |
BC095496.1 | 1 | 576 | AAH95496.1 | |
BM671230.1 | 1 | 505 | ||
BX537430.1 | 3 | 1417 | CAD97672.1 | |
CA435450.1 | 1 | 505 | ||
EL735542.1 | ||||
HE572751.1 | 2 | 573 | ||
JX081310.1 | 1 | 579 | ||
U01839.1 | 1 | 754 | AAC50055.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)