Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | NP_001139287.1 | ||
NM_015944.3 | NP_057028.2 | |||
XM_017023262.1 | XP_016878751.1 | |||
XM_017023263.1 | XP_016878752.1 | |||
XM_017023264.1 | XP_016878753.1 | |||
XM_017023265.1 | XP_016878754.1 | |||
XM_017023266.1 | XP_016878755.1 | |||
XM_017023267.1 | XP_016878756.1 | |||
AB209351.1 | BAD92588.1 | |||
AF132948.1 | AAD27723.1 | |||
AK296877.1 | ||||
AK300747.1 | ||||
BC018734.1 | AAH18734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | C1QTNF8 NME3 LOC652276 LOC101929280 RPL23AP5 FAM234A HBA1 HN1L MIR3176 MAPK8IP3 AXIN1 BRICD5 DNASE1L2 HS3ST6 TELO2 ZNF598 LOC105371045 TRAF7 MIR5587 MRPS34 MIR940 MIR3677 PDPK1 E4F1 GNG13 TPSB2 CCNF DDX11L10 MIR3180-5 MRPL28 SLC9A3R2 MIR662 FAHD1 CEMP1 PDIA2 IFT140 MIR4516 MLST8 HAGH FLJ42627 PRR35 PRSS29P SNRNP25 PTX4 MIR6511B1 SNHG19 C16orf91 NPW CACNA1H RAB26 TSC2 SSTR5 MIR6767 KCTD5 ECI1 LOC105371184 GNPTG ABCA3 PRSS27 METRN UBE2I MIR3177 LOC729652 MSRB1 RGS11 GFER LINC00235 TBL3 NTHL1 NUBP2 SPSB3 RNPS1 CCDC154 FAM173A LINC00254 RHBDF1 LOC100134368 SOX8 CAPN15 TMEM204 SSTR5-AS1 TSR3 LOC107987233 LOC106660606 METTL26 PGP ATP6V0C NTN3 IGFALS RPL3L HBZ NARFL SNORA64 EME2 MIR6768 C16orf59 JMJD8 HBQ1 LOC105371046 RPUSD1 NHLRC4 LUC7L AMDHD2 SNORA78 LOC105371047 RAB40C MIR3178 LOC100287175 NDUFB10 WASIR2 POLR3K RPS2 RHBDL1 DECR2 HAGHL MIR6859-4 LOC107984876 TPSG1 TPSAB1 ABCA17P SYNGR3 FBXL16 UNKL CCDC78 SNORD60 MIR4717 NPRL3 RNF151 RHOT2 MEIOB MIR1225 TPSD1 SNHG9 CLCN7 ERVK13-1 ARHGDIG LMF1 PKD1 MSLN CASKIN1 STUB1 NME4 MPG LMF1-AS1 LOC105371038 BAIAP3 CHTF18 HBM PIGQ NOXO1 RAB11FIP3 WDR90 TBC1D24 TMEM8A CRAMP1 WFIKKN1 MCRIP2 PRR25 HBA2 WDR24 SNORA10 LOC105371049 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | TBC1D24 MIR3178 SRRM2-AS1 PRSS33 LOC652276 CEMP1 SRRM2 FLJ42627 TCEB2 PDPK1 ERVK13-1 PRSS21 AMDHD2 KCTD5 PRSS41 PRSS27 ATP6V0C |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | TBC1D24 MIR3178 AMDHD2 LOC652276 CEMP1 FLJ42627 PDPK1 ERVK13-1 ATP6V0C |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | SNORD60 NTN3 MIR4717 LOC652276 MIR6768 MIR1225 CEMP1 MIR4516 C16orf59 MLST8 BRICD5 DNASE1L2 AMDHD2 PKD1 MIR6511B1 SNHG19 CASKIN1 RAB26 TSC2 MIR6767 ECI1 TRAF7 ABCA3 TBC1D24 MIR3178 LOC729652 MIR940 MIR3677 PDPK1 RNPS1 E4F1 ABCA17P LOC105371049 CCNF MIR3180-5 LOC106660606 PGP ATP6V0C |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |