Assay Details
Target Gene Details
Entrez Gene ID: | 56479 |
Gene Name: | potassium voltage-gated channel subfamily Q member 5 |
Gene Aliases: |
Kv7.5 |
Location: |
Chr.6:72621843-73198851 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNQ5 | NM_001160130.1 | 1 | 672 | NP_001153602.1 |
NM_001160132.1 | 1 | 672 | NP_001153604.1 | |
NM_001160133.1 | 1 | 672 | NP_001153605.1 | |
NM_001160134.1 | 1 | 672 | NP_001153606.1 | |
NM_019842.3 | 1 | 672 | NP_062816.2 | |
XM_017011058.1 | 1 | 672 | XP_016866547.1 | |
AF202977.1 | 1 | 220 | AAF69797.1 | |
AF249278.1 | 1 | 434 | AAF91335.1 | |
AF263835.1 | 1 | 112 | AAF73446.1 | |
AK299550.1 | 1 | 272 | ||
BC050689.1 | 1 | 409 | AAH50689.1 | |
BC117359.1 | 1 | 343 | ||
BC143554.1 | 1 | 343 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |