Assay Details
Target Gene Details
Entrez Gene ID: | 9049 |
Gene Name: | aryl hydrocarbon receptor interacting protein |
Gene Aliases: |
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2 |
Location: |
Chr.11:67483029-67491108 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIP | NM_001302960.1 | NP_001289889.1 | ||
NM_003977.3 | NP_003968.3 | |||
BC104797.1 | AAI04798.1 | |||
BC104827.1 | AAI04828.1 | |||
BG821308.1 | ||||
HY065637.1 | ||||
U31913.1 | AAB39923.1 | |||
U78521.1 | AAB59004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv368 | Chr.11:67479842 - 67509866 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv825960 | Chr.11:67471173 - 67483362 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)