Assay Details
Target Gene Details
Entrez Gene ID: | 2804 |
Gene Name: | golgin B1 |
Gene Aliases: |
GCP, GCP372, GOLIM1 |
Location: |
Chr.3:121663199-121749908 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8524n54 | Chr.3:121252053 - 121838686 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829703 | Chr.3:121664443 - 121815646 on Build GRCh38 | Loss |
![]() ![]() |
esv3575599 | Chr.3:121571453 - 121671424 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv998740 | Chr.3:121570652 - 121671424 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3575598 | Chr.3:121406926 - 121715587 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv999335 | Chr.3:121381436 - 121689919 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)