Assay Details
Target Gene Details
Entrez Gene ID: | 8786 |
Gene Name: | regulator of G-protein signaling 11 |
Gene Aliases: |
RS11 |
Location: |
Chr.16:268301-275943 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS11 | NM_001286485.1 | NP_001273414.1 | ||
NM_001286486.1 | NP_001273415.1 | |||
NM_003834.2 | NP_003825.1 | |||
NM_183337.2 | NP_899180.1 | |||
XM_011522719.2 | XP_011521021.1 | |||
XM_011522720.2 | XP_011521022.1 | |||
AB016929.1 | BAA74751.1 | |||
AB209582.1 | BAD92819.1 | |||
AF035153.1 | AAC69175.1 | |||
AF035154.1 | AAC69176.1 | |||
AK091701.1 | ||||
AK293564.1 | ||||
BC141803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | SNHG9 IFT140 MIR6511B1 LMF1-AS1 NOXO1 TPSD1 WDR24 SSTR5 LINC00254 MLST8 SNORA10 EME2 MAPK8IP3 PKD1 RAB40C HS3ST6 NTHL1 NME4 BAIAP3 CCDC78 PRR35 NARFL MIR3180-5 HAGH MIR6859-4 LOC729652 SNRNP25 MIR940 TSC2 PRSS29P LOC105371045 SSTR5-AS1 MIR5587 JMJD8 MIR662 HBQ1 LOC652276 BRICD5 MIR6767 WDR90 MIR3177 METTL26 DDX11L10 PRSS27 SNORA78 TELO2 SLC9A3R2 LOC107984876 TBL3 RNF151 ECI1 PIGQ FAHD1 CASKIN1 MSLN PTX4 SOX8 HBZ MIR3176 NUBP2 C16orf91 LOC105371038 LUC7L DNASE1L2 TPSB2 ABCA3 CHTF18 CEMP1 RHOT2 HBA1 RGS11 SNHG19 TRAF7 MRPS34 LOC106660606 LOC105371047 RPL23AP5 SNORD60 RHBDL1 SPSB3 MEIOB POLR3K CAPN15 E4F1 NPW MIR4717 RPS2 PDIA2 CCNF ABCA17P SYNGR3 MIR4516 TMEM8A DECR2 WASIR2 WFIKKN1 NDUFB10 HBM CLCN7 LOC100134368 C16orf59 GNG13 ZNF598 TMEM204 PGP UBE2I CACNA1H FBXL16 HAGHL HN1L RPUSD1 NTN3 IGFALS KCTD5 NPRL3 RAB11FIP3 PDPK1 LINC00235 ARHGDIG CRAMP1 RAB26 LOC105371046 LMF1 ATP6V0C SNORA64 MIR3677 FAM173A TPSAB1 AXIN1 MIR1225 TSR3 MRPL28 METRN LOC105371049 LOC100287175 TPSG1 RNPS1 LOC101929280 C1QTNF8 GNPTG NME3 HBA2 ERVK13-1 MPG GFER FAM234A AMDHD2 MIR6768 PRR25 RHBDF1 FLJ42627 MCRIP2 RPL3L TBC1D24 CCDC154 LOC105371184 UNKL STUB1 MIR3178 MSRB1 NHLRC4 LOC107987233 |
esv3570031 | Chr.16:243389 - 278107 on Build GRCh38 | Gain | FAM234A RGS11 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | ARHGDIG MRPL28 RPL23AP5 LOC100134368 PDIA2 FAM234A RGS11 TMEM8A DECR2 AXIN1 NME4 RAB11FIP3 |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain | ARHGDIG PDIA2 FAM234A RGS11 AXIN1 |
esv3637561 | Chr.16:261769 - 271795 on Build GRCh38 | Loss | FAM234A RGS11 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | ARHGDIG MRPL28 RPL23AP5 LOC100134368 PDIA2 FAM234A RGS11 TMEM8A DECR2 AXIN1 NME4 RAB11FIP3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |