Assay Details
Target Gene Details
Entrez Gene ID: | 285440 |
Gene Name: | cytochrome P450 family 4 subfamily V member 2 |
Gene Aliases: |
BCD, CYP4AH1 |
Location: |
Chr.4:186191520-186213463 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP4V2 | NM_207352.3 | 11 | 4382 | NP_997235.3 |
XM_005262935.3 | 11 | 4164 | XP_005262992.1 | |
XM_017008037.1 | 10 | 4140 | XP_016863526.1 | |
AA918796.1 | 1 | 332 | ||
AK126473.1 | 12 | 4051 | BAC86562.1 | |
BX648537.1 | 5 | 3313 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain | FAM149A TLR3 FLJ38576 KLKB1 SORBS2 F11 CYP4V2 F11-AS1 |
nsv4644 | Chr.4:186185600 - 186219291 on Build GRCh38 | Insertion | FLJ38576 KLKB1 CYP4V2 |
nsv830172 | Chr.4:186091611 - 186247270 on Build GRCh38 | Loss | FAM149A FLJ38576 KLKB1 CYP4V2 |
nsv508340 | Chr.4:186138019 - 186215804 on Build GRCh38 | Deletion | FAM149A FLJ38576 KLKB1 CYP4V2 |
esv3603523 | Chr.4:186176579 - 186214960 on Build GRCh38 | Gain | FLJ38576 CYP4V2 |
nsv461864 | Chr.4:186085652 - 186335850 on Build GRCh38 | Gain | FAM149A TLR3 FLJ38576 KLKB1 F11 CYP4V2 F11-AS1 |
nsv596401 | Chr.4:186151229 - 186445495 on Build GRCh38 | Loss | FAM149A FLJ38576 KLKB1 F11 CYP4V2 F11-AS1 |
nsv4642 | Chr.4:186164028 - 186215260 on Build GRCh38 | Deletion | FAM149A FLJ38576 CYP4V2 |
esv3603525 | Chr.4:186210472 - 186225457 on Build GRCh38 | Loss | KLKB1 CYP4V2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |