Assay Details
Target Gene Details
Entrez Gene ID: | 51422 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 2 |
Gene Aliases: |
AAKG, AAKG2, CMH6, H91620p, WPWS |
Location: |
Chr.7:151556114-151877231 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831189 | Chr.7:151452021 - 151622168 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)