Assay Details
Target Gene Details
Entrez Gene ID: | 81502 |
Gene Name: | histocompatibility minor 13 |
Gene Aliases: |
H13, IMP1, IMPAS, IMPAS-1, MSTP086, PSENL3, PSL3, SPP, SPPL1 |
Location: |
Chr.20:31514410-31569567 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HM13 | NM_030789.3 | 12 | 1223 | NP_110416.1 |
NM_178580.2 | 13 | 1426 | NP_848695.1 | |
NM_178581.2 | 13 | 1370 | NP_848696.1 | |
AF172086.1 | 12 | 1171 | AAQ13609.1 | |
AF483215.1 | 12 | 1169 | AAM22076.1 | |
AF515663.1 | AAN77099.1 | |||
AJ345029.1 | 12 | 1166 | CAC87790.1 | |
AJ420895.1 | 12 | 1105 | CAD13132.1 | |
AK075283.1 | 12 | 1183 | BAC11519.1 | |
AK314410.1 | ||||
AY169310.1 | 13 | 1383 | AAO12535.1 | |
AY169311.1 | 12 | 1180 | AAO12536.1 | |
AY169312.1 | 13 | 1327 | AAO12537.1 | |
BC008938.2 | 12 | 1194 | AAH08938.1 | |
BC008959.2 | 12 | 1194 | AAH08959.1 | |
BC062595.1 | 12 | 1174 | AAH62595.1 | |
DQ168450.1 | 13 | 1378 | ||
KU178790.1 | ||||
KU178791.1 |
Target Gene Details
Entrez Gene ID: | 100874042 |
Gene Name: | HM13 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:31567587-31573263 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HM13-AS1 | NR_046853.2 | |||
AJ710081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470546 | Chr.20:31245806 - 31647499 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2763228 | Chr.20:31247246 - 31651719 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3645652 | Chr.20:31500710 - 31576564 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv3345 | Chr.20:31549053 - 31593946 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv458961 | Chr.20:31305698 - 31688103 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)