Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BT006664.1 | AAP35310.1 | |||
CR457404.1 | CAG33685.1 | |||
D13138.1 | BAA02431.1 | |||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | TCF25 VPS9D1 LOC107984817 CHMP1A AFG3L1P LOC105371419 MC1R VPS9D1-AS1 TUBB3 SPATA2L DEF8 DPEP1 GAS8-AS1 SPATA33 CDK10 SPIRE2 DBNDD1 CENPBD1 GAS8 ZNF276 FANCA |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv1930 | Chr.16:89633876 - 89640750 on Build GRCh38 | Insertion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | TUBB8P7 VPS9D1 CHMP1A SNORD68 VPS9D1-AS1 TUBB3 LOC105371414 SPATA2L RPL13 SPG7 GAS8-AS1 CDK10 LOC101927817 PRDM7 SPIRE2 DBNDD1 CENPBD1 LOC100287036 GAS8 ZNF276 TCF25 FAM157C ZNF778 LOC107984817 AFG3L1P LOC105371419 MC1R DEF8 DPEP1 CPNE7 SPATA33 ANKRD11 URAHP FANCA |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | SPATA2L DPEP1 CHMP1A SPATA33 CDK10 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | RPL13 DPEP1 CPNE7 SPG7 CHMP1A SPATA33 LOC101927817 ANKRD11 SNORD68 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | VPS9D1 LOC339059 LOC107987238 VPS9D1-AS1 TUBB3 SPATA2L RPL13 LOC100289580 GAS8-AS1 PRDM7 CTU2 SPIRE2 LOC100287036 SLC22A31 GAS8 ZNF276 LOC101927793 CDT1 PIEZO1 CBFA2T3 TCF25 CYBA FAM157C ZNF778 CDH15 DPEP1 LOC400558 CPNE7 SNAI3 SPATA33 TRAPPC2L PABPN1L ANKRD11 URAHP FANCA TUBB8P7 RNF166 CHMP1A LOC105371409 LINC00304 MIR4722 LOC100129697 SNORD68 LOC105371414 SPG7 GALNS CDK10 ACSF3 LOC101927817 DBNDD1 CENPBD1 IL17C APRT LOC107984817 AFG3L1P LOC105371419 MC1R DEF8 MVD SNAI3-AS1 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | VPS9D1 CHMP1A SNORD68 VPS9D1-AS1 SPATA2L RPL13 DPEP1 CPNE7 SPG7 SPATA33 CDK10 ZNF276 FANCA |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | RPL13 DPEP1 CPNE7 SPG7 CHMP1A SPATA33 SNORD68 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |