Assay Details
Target Gene Details
Entrez Gene ID: | 56910 |
Gene Name: | StAR related lipid transfer domain containing 7 |
Gene Aliases: |
GTT1 |
Location: |
Chr.2:96184859-96208846 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7 | NM_020151.3 | 8 | 3234 | NP_064536.2 |
XM_011511499.1 | 8 | 2851 | XP_011509801.1 | |
AB075479.1 | 1 | 860 | ||
AF270647.1 | 8 | 3161 | AAF81750.1 | |
AK023234.1 | 7 | 2493 | ||
AK054615.1 | 8 | 2600 | ||
AK094925.1 | 2 | 2806 | ||
AK096666.1 | 5 | 1994 | ||
BC007894.2 | 9 | 2992 | AAH07894.1 | |
BC008894.2 | 8 | 3217 | AAH08894.1 | |
BC009998.2 | 9 | 2992 | AAH09998.1 | |
BC012774.2 | 8 | 3184 | AAH12774.1 | |
BC012793.2 | 8 | 3198 | AAH12793.1 | |
BC013279.2 | 8 | 3217 | AAH13279.1 | |
BC014076.2 | 8 | 3194 | AAH14076.1 | |
BC014274.2 | 8 | 3213 | AAH14274.3 | |
BC032106.1 | 8 | 3208 | AAH32106.1 | |
CR749695.1 | 1 | 1549 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | STARD7-AS1 CIAO1 TMEM127 LOC105373495 ITPRIPL1 STARD7 SNRNP200 NCAPH |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | FAHD2CP ADRA2B LOC107984110 LOC100506076 TMEM127 ARID5A ITPRIPL1 KANSL3 CNNM4 MIR3127 SEMA4C ANKRD39 CIAO1 FAHD2B ANKRD36B STARD7 ANKRD23 ANKRD36 CNNM3 LOC653924 LMAN2L NEURL3 LOC101927053 DUSP2 STARD7-AS1 FAM178B ASTL ANKRD36C FER1L5 LOC105373495 SNRNP200 NCAPH LOC105373496 LOC100506123 GPAT2 |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | FAHD2CP ADRA2B LOC107984110 LOC100506076 TMEM127 ARID5A ITPRIPL1 KANSL3 CNNM4 MIR3127 SEMA4C ANKRD39 CIAO1 FAHD2B ANKRD36B STARD7 ANKRD23 ANKRD36 CNNM3 LOC653924 LMAN2L NEURL3 LOC101927053 DUSP2 STARD7-AS1 FAM178B ASTL ANKRD36C FER1L5 LOC105373495 SNRNP200 NCAPH LOC105373496 LOC100506123 GPAT2 |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | FAHD2CP ADRA2B LOC107984110 LOC100506076 TMEM127 ARID5A ITPRIPL1 KANSL3 CNNM4 MIR3127 SEMA4C ANKRD39 CIAO1 FAHD2B ANKRD36B STARD7 ANKRD23 ANKRD36 CNNM3 LOC653924 LMAN2L NEURL3 LOC101927053 DUSP2 STARD7-AS1 FAM178B ASTL ANKRD36C FER1L5 LOC105373495 SNRNP200 NCAPH LOC105373496 LOC100506123 GPAT2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |