Assay Details
Target Gene Details
Entrez Gene ID: | 26268 |
Gene Name: | F-box protein 9 |
Gene Aliases: |
FBX9, NY-REN-57, VCIA1, dJ341E18.2 |
Location: |
Chr.6:53057807-53100873 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXO9 | NM_012347.4 | 4 | 453 | NP_036479.1 |
NM_033480.2 | 5 | 703 | NP_258441.1 | |
NM_033481.3 | 5 | 580 | NP_258442.2 | |
XM_005248995.4 | 5 | 387 | XP_005249052.1 | |
XM_005248996.4 | 5 | 538 | XP_005249053.1 | |
AB209833.1 | 5 | 742 | BAD93070.1 | |
AF174597.1 | AAF04518.1 | |||
AF176704.1 | 4 | 453 | AAF03704.1 | |
AK021559.1 | 5 | 739 | ||
AL137520.1 | 5 | 703 | CAB70786.2 | |
AY858798.1 | 5 | 745 | AAW51115.1 | |
BC000650.2 | 5 | 580 | AAH00650.2 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |