Assay Details
Target Gene Details
Entrez Gene ID: | 84435 |
Gene Name: | adhesion G protein-coupled receptor A1 |
Gene Aliases: |
GPR123 |
Location: |
Chr.10:133070929-133131675 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRA1 | XM_005252695.1 | 8 | 1493 | XP_005252752.1 |
AY181241.1 | 8 | 1493 | AAO27353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759793 | Chr.10:133030918 - 133619181 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv998n100 | Chr.10:132998894 - 133170978 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3625022 | Chr.10:132998655 - 133120915 on Build GRCh38 | Gain |
![]() ![]() |
nsv467498 | Chr.10:132900981 - 133134139 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3625015 | Chr.10:132889012 - 133114687 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv528260 | Chr.10:132859348 - 133620799 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952703 | Chr.10:133037897 - 133143896 on Build GRCh38 | Deletion |
![]() ![]() |
nsv470979 | Chr.10:133033478 - 133118280 on Build GRCh38 | Loss |
![]() ![]() |
nsv552526 | Chr.10:132699017 - 133103793 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3625027 | Chr.10:133082541 - 133084963 on Build GRCh38 | Loss |
![]() |
nsv552539 | Chr.10:133061918 - 133103793 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs114900682] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)