Assay Details
Target Gene Details
Entrez Gene ID: | 123803 |
Gene Name: | N-terminal asparagine amidase |
Gene Aliases: |
PNAA, PNAD |
Location: |
Chr.16:15037853-15056079 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTAN1 | NM_001270766.1 | NP_001257695.1 | ||
NM_001270767.1 | NP_001257696.1 | |||
NM_173474.3 | NP_775745.1 | |||
AF092440.1 | AAP97215.1 | |||
BC017336.2 | AAH17336.1 | |||
BF939373.1 | ||||
DC383993.1 | ||||
HY036316.1 |
Target Gene Details
Entrez Gene ID: | 23042 |
Gene Name: | pyridoxal dependent decarboxylase domain containing 1 |
Gene Aliases: |
LP8165 |
Location: |
Chr.16:14974591-15139410 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDXDC1 | NM_001285449.1 | NP_001272378.1 | ||
NM_001324020.1 | NP_001310949.1 | |||
NM_001324021.1 | NP_001310950.1 | |||
XM_017023065.1 | XP_016878554.1 | |||
AY203955.1 | AAP34478.1 | |||
BC045554.1 | AAH45554.2 | |||
CD370048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638022 | Chr.16:15031773 - 15047500 on Build GRCh38 | Loss | PDXDC1 NTAN1 |
dgv4922n54 | Chr.16:15025750 - 15099845 on Build GRCh38 | Loss | PDXDC1 LOC100505915 RRN3 NTAN1 |
esv3570354 | Chr.16:15034129 - 15223463 on Build GRCh38 | Gain | PDXDC1 PKD1P6-NPIPP1 LOC100505915 RRN3 MIR6511B2 MIR3180-4 NTAN1 |
nsv1050325 | Chr.16:14954899 - 15166867 on Build GRCh38 | Loss | LOC728138 PDXDC1 PKD1P6-NPIPP1 LOC100505915 RRN3 MIR6511B2 MIR3180-4 MIR1972-1 NTAN1 |
nsv509598 | Chr.16:15025400 - 15067337 on Build GRCh38 | Insertion | PDXDC1 RRN3 NTAN1 |
nsv1038955 | Chr.16:14885100 - 15322641 on Build GRCh38 | Loss | LOC728138 NOMO1 LOC100505915 MIR6770-1 MIR1972-1 MIR3179-1 NTAN1 LOC100288162 PDXDC1 PKD1P6-NPIPP1 RRN3 MIR6511B2 MIR3180-4 MIR3670-1 MIR6511A1 MIR3180-1 NPIPA1 |
esv2758632 | Chr.16:14528198 - 15272693 on Build GRCh38 | Gain+Loss | NOMO1 BFAR LOC105376747 MIR6770-1 MIR1972-1 NPIPA3 MIR3179-1 NTAN1 LOC100652777 PKD1P6-NPIPP1 ABCC6P2 MIR3670-1 NPIPA2 NPIPA1 LOC728138 LOC100505915 LOC100288162 PARN PDXDC1 RRN3 MIR6511B2 MIR3180-4 PLA2G10 MIR6511A1 MIR3180-1 LOC105371095 |
esv2762254 | Chr.16:14741210 - 15322653 on Build GRCh38 | Gain+Loss | LOC728138 NOMO1 LOC100505915 LOC105376747 MIR6770-1 MIR1972-1 MIR3179-1 NTAN1 LOC100288162 PDXDC1 PKD1P6-NPIPP1 RRN3 MIR6511B2 MIR3180-4 ABCC6P2 MIR3670-1 MIR6511A1 MIR3180-1 NPIPA2 NPIPA1 |
dgv2755n100 | Chr.16:14877546 - 15434384 on Build GRCh38 | Gain | LOC728138 NOMO1 LOC100505915 MIR6770-1 NPIPA5 MIR1972-1 MIR3179-1 NTAN1 LOC100288162 PDXDC1 PKD1P6-NPIPP1 RRN3 MIR6511B2 MIR3180-4 MIR3670-1 MPV17L MIR6511A1 MIR3180-1 NPIPA1 |
esv3892796 | Chr.16:14961008 - 15074903 on Build GRCh38 | Loss | LOC728138 PDXDC1 RRN3 MIR1972-1 NTAN1 |
nsv1053467 | Chr.16:15031584 - 15322641 on Build GRCh38 | Loss | PDXDC1 PKD1P6-NPIPP1 LOC100505915 RRN3 MIR6511B2 MIR3180-4 NTAN1 |
esv3638020 | Chr.16:15031082 - 15082591 on Build GRCh38 | Loss | PDXDC1 RRN3 NTAN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |