Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | 1 | 70 | NP_057356.1 |
XM_005261315.2 | 1 | 1277 | XP_005261372.1 | |
XM_017028539.1 | 1 | 1277 | XP_016884028.1 | |
BM453650.1 | 1 | 70 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss | RANGAP1 LOC100996598 ZC3H7B TOB2 TEF PHF5A |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss | LOC105373044 CSDC2 PMM1 ACO2 RANGAP1 LOC100996598 ZC3H7B TOB2 TEF POLR3H PHF5A MIR6889 |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss | LOC105373044 ACO2 LOC100996598 TOB2 TEF POLR3H PHF5A |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain | RANGAP1 LOC100996598 ZC3H7B TOB2 TEF MIR6889 CHADL |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | LOC105373044 C22orf46 PMM1 RANGAP1 ZC3H7B TEF EP300 L3MBTL2 PHF5A MIR6889 EP300-AS1 DESI1 CSDC2 MIR1281 ACO2 LOC100996598 TOB2 XRCC6 POLR3H MEI1 SNORD140 CHADL SNU13 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |