Assay Details
Target Gene Details
Entrez Gene ID: | 79140 |
Gene Name: | coiled-coil domain containing 28B |
Gene Aliases: |
- |
Location: |
Chr.1:32200386-32205390 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC28B | NM_001301011.1 | 2 | 458 | NP_001287940.1 |
NM_024296.4 | 2 | 458 | NP_077272.2 | |
XM_011542113.2 | 2 | 1217 | XP_011540415.1 | |
XM_017002306.1 | 2 | 458 | XP_016857795.1 | |
XM_017002307.1 | 2 | 1212 | XP_016857796.1 | |
XM_017002308.1 | 2 | 458 | XP_016857797.1 | |
XM_017002309.1 | 2 | 1215 | XP_016857798.1 | |
AK092702.1 | 2 | 244 | ||
AK225113.1 | 2 | 228 | ||
AK291904.1 | 2 | 468 | ||
BC022848.2 | 2 | 192 | AAH22848.1 | |
BM981930.1 | ||||
DA899719.1 | 2 | 244 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |