Assay Details
Target Gene Details
Entrez Gene ID: | 6354 |
Gene Name: | C-C motif chemokine ligand 7 |
Gene Aliases: |
FIC, MARC, MCP-3, MCP3, NC28, SCYA6, SCYA7 |
Location: |
Chr.17:34270216-34272242 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL7 | NM_006273.3 | 3 | 353 | NP_006264.2 |
AF043338.1 | AAC03538.1 | |||
BC092436.1 | 3 | 347 | AAH92436.1 | |
BC112258.1 | 3 | 343 | AAI12259.1 | |
BC112260.1 | 3 | 343 | AAI12261.1 | |
CD523198.1 | 3 | 356 | ||
X71087.1 | 3 | 348 | CAA50405.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3141n100 | Chr.17:33630132 - 34611003 on Build GRCh38 | Gain | ASIC2 CCL11 C17orf102 LOC107987247 LOC101927239 CCL2 CCL13 CCL7 TMEM132E CCL1 CCL8 |
nsv517423 | Chr.17:33777656 - 34498242 on Build GRCh38 | Gain+Loss | ASIC2 CCL11 LOC107987247 LOC101927239 CCL2 CCL13 CCL7 CCL1 CCL8 |
nsv574714 | Chr.17:33640441 - 34608994 on Build GRCh38 | Gain | ASIC2 CCL11 C17orf102 LOC107987247 LOC101927239 CCL2 CCL13 CCL7 TMEM132E CCL1 CCL8 |
esv3572553 | Chr.17:33632208 - 34617133 on Build GRCh38 | Gain | ASIC2 CCL11 C17orf102 LOC107987247 LOC101927239 CCL2 CCL13 CCL7 TMEM132E CCL1 CCL8 |
nsv519297 | Chr.17:33640660 - 34608994 on Build GRCh38 | Gain | ASIC2 CCL11 C17orf102 LOC107987247 LOC101927239 CCL2 CCL13 CCL7 TMEM132E CCL1 CCL8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |