Assay Details
Target Gene Details
Entrez Gene ID: | 1632 |
Gene Name: | enoyl-CoA delta isomerase 1 |
Gene Aliases: |
DCI |
Location: |
Chr.16:2239872-2251601 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECI1 | NM_001178029.1 | NP_001171500.1 | ||
NM_001919.3 | NP_001910.2 | |||
AK291127.1 | ||||
BC000762.2 | AAH00762.1 | |||
BC002746.2 | AAH02746.1 | |||
BC009631.1 | AAH09631.1 | |||
BC019316.1 | AAH19316.1 | |||
BC020228.1 | AAH20228.1 | |||
BQ773923.1 | ||||
CD675473.1 | ||||
L24774.1 | AAA35485.1 | |||
Z25820.1 | CAA81065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571200 | Chr.16:2209190 - 2244889 on Build GRCh38 | Loss | BRICD5 E4F1 DNASE1L2 PGP MLST8 ECI1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | LMF1-AS1 MIR3177 SYNGR3 PDIA2 SNRNP25 SOX8 TMEM204 SNHG9 ZNF598 MCRIP2 MIR662 RAB11FIP3 RPUSD1 RPL3L PRSS27 RHOT2 MAPK8IP3 RGS11 NPW LOC107987233 LOC105371049 C1QTNF8 LOC105371184 LOC101929280 EME2 GNPTG FLJ42627 NHLRC4 MIR3176 MPG PIGQ AXIN1 MIR3677 LOC105371047 SPSB3 CAPN15 NME3 HBZ WDR24 SNHG19 MRPS34 LOC729652 MRPL28 MIR3180-5 MIR4516 C16orf59 TPSB2 IFT140 BRICD5 FBXL16 CCDC154 KCTD5 CACNA1H AMDHD2 ARHGDIG ATP6V0C RPS2 ABCA3 RNF151 PRSS29P DDX11L10 CLCN7 WFIKKN1 MIR6767 SNORD60 PGP LOC652276 HAGH UBE2I STUB1 LOC100287175 PRR35 SNORA10 ABCA17P TPSG1 TMEM8A TPSD1 NUBP2 PDPK1 CCDC78 LINC00254 SNORA64 RHBDL1 LUC7L LOC106660606 NARFL MLST8 RAB40C LOC105371046 GNG13 FAM234A DECR2 NME4 METRN SSTR5 HBQ1 SLC9A3R2 NPRL3 ECI1 MIR1225 TELO2 SNORA78 SSTR5-AS1 NDUFB10 CRAMP1 RAB26 POLR3K METTL26 MSRB1 HBA2 CEMP1 TBC1D24 HN1L MIR940 FAM173A UNKL LOC105371045 LOC105371038 MEIOB NTN3 WASIR2 CASKIN1 FAHD1 NTHL1 E4F1 HS3ST6 CCNF RNPS1 ERVK13-1 RHBDF1 HAGHL LINC00235 TSR3 PTX4 TBL3 TSC2 MIR6859-4 WDR90 TPSAB1 MIR6511B1 CHTF18 PRR25 TRAF7 MIR6768 LOC100134368 PKD1 LOC107984876 DNASE1L2 C16orf91 HBM RPL23AP5 JMJD8 MIR4717 LMF1 NOXO1 HBA1 MIR3178 MIR5587 IGFALS BAIAP3 MSLN GFER |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | CASKIN1 SYNGR3 NTHL1 E4F1 SNHG19 LOC106660606 MLST8 RNPS1 MIR3180-5 MIR4516 ZNF598 BRICD5 TBL3 TSC2 MIR6511B1 SLC9A3R2 ECI1 TRAF7 NPW PKD1 MIR1225 ABCA3 DNASE1L2 LOC105371049 RAB26 MIR4717 NOXO1 SNORD60 PGP MIR940 MIR3677 GFER |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | TRAF7 CASKIN1 PKD1 MIR1225 E4F1 DNASE1L2 LOC105371049 SNHG19 MLST8 RAB26 MIR3180-5 MIR4516 BRICD5 TSC2 SNORD60 MIR6511B1 PGP ECI1 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | TRAF7 CASKIN1 E4F1 DNASE1L2 SNHG19 LOC106660606 MLST8 RAB26 RNPS1 BRICD5 SNORD60 PGP MIR940 ECI1 MIR3677 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | TRAF7 CASKIN1 BRICD5 E4F1 DNASE1L2 PGP LOC106660606 MIR940 MLST8 RNPS1 ECI1 MIR3677 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | ABCA17P PDPK1 CASKIN1 E4F1 SNHG19 CCNF LOC106660606 MLST8 RNPS1 LOC729652 MIR3180-5 MIR4516 C16orf59 BRICD5 TSC2 MIR6511B1 ECI1 AMDHD2 ATP6V0C TRAF7 MIR6768 PKD1 MIR1225 ABCA3 DNASE1L2 LOC105371049 RAB26 MIR4717 CEMP1 MIR3178 TBC1D24 MIR6767 SNORD60 PGP MIR940 LOC652276 MIR3677 NTN3 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | TRAF7 CASKIN1 E4F1 ABCA3 DNASE1L2 SNHG19 LOC106660606 MLST8 RAB26 RNPS1 MIR4717 BRICD5 SNORD60 PGP MIR940 ECI1 MIR3677 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |