Assay Details
Target Gene Details
Entrez Gene ID: | 121260 |
Gene Name: | solute carrier family 15 member 4 |
Gene Aliases: |
FP12591, PHT1, PTR4 |
Location: |
Chr.12:128793194-128823996 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC15A4 | NM_145648.3 | 5 | 1188 | NP_663623.1 |
XM_011537895.1 | 5 | 1308 | XP_011536197.1 | |
XM_017018791.1 | 5 | 1308 | XP_016874280.1 | |
XM_017018792.1 | 5 | 1308 | XP_016874281.1 | |
XM_017018793.1 | 5 | 1158 | XP_016874282.1 | |
AF461893.1 | 3 | 310 | AAQ04807.1 | |
AK095717.1 | 4 | 802 | ||
AY038999.1 | 5 | 1193 | AAK72099.1 | |
AY050629.1 | 5 | 1138 | AAK95565.1 | |
BC028394.2 | 5 | 941 | AAH28394.2 | |
BC146803.1 | 5 | 1164 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2963n54 | Chr.12:128794719 - 128847110 on Build GRCh38 | Loss | SLC15A4 LOC101929984 |
nsv1051433 | Chr.12:128679360 - 128888555 on Build GRCh38 | Gain | LOC100128276 GLT1D1 SLC15A4 TMEM132C LOC101929984 |
esv3631151 | Chr.12:128793327 - 128853915 on Build GRCh38 | Gain | LOC100128276 GLT1D1 SLC15A4 LOC101929984 |
esv2759920 | Chr.12:128587760 - 128896765 on Build GRCh38 | Loss | LOC100128276 GLT1D1 SLC15A4 TMEM132C LOC101929984 |
dgv528e212 | Chr.12:128795042 - 128814506 on Build GRCh38 | Loss | SLC15A4 |
nsv1040831 | Chr.12:128749443 - 129516029 on Build GRCh38 | Gain | LOC101927735 LOC100128276 GLT1D1 SLC15A4 LOC283352 TMEM132D LOC101929984 |
nsv832549 | Chr.12:128739165 - 128936554 on Build GRCh38 | Loss | LOC100128276 GLT1D1 SLC15A4 LOC101929984 |
esv3581993 | Chr.12:128679608 - 128885566 on Build GRCh38 | Gain | LOC100128276 GLT1D1 SLC15A4 TMEM132C LOC101929984 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |