Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | BAC04000.1 | |||
AK292004.1 | ||||
AK300915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | EXOC3L1 DUS2 NOL3 LOC105369155 LOC100131303 LRRC29 E2F4 LOC107984813 ELMO3 ACD DDX28 TMEM208 MIR328 TRADD PSKH1 NRN1L DPEP2 THAP11 PSMB10 ATP6V0D1 EDC4 CTRL NUTF2 KIAA0895L KCTD19 HSF4 TSNAXIP1 TPPP3 PARD6A FAM65A SLC9A5 DPEP3 GFOD2 FBXL8 CTCF AGRP ENKD1 LRRC36 SLC12A4 LCAT ZDHHC1 CARMIL2 FHOD1 CENPT HSD11B2 PLEKHG4 C16orf86 LOC100505942 RANBP10 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | MIR328 TRADD EXOC3L1 SLC9A5 NOL3 C16orf70 B3GNT9 FBXL8 LOC105369155 LRRC29 E2F4 FHOD1 KIAA0895L HSF4 PLEKHG4 ELMO3 TMEM208 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | MIR328 TRADD EXOC3L1 NOL3 C16orf70 B3GNT9 FBXL8 LOC105369155 LRRC29 E2F4 FHOD1 KIAA0895L HSF4 ELMO3 TMEM208 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | MIR328 EXOC3L1 LRRC29 E2F4 NOL3 KIAA0895L HSF4 FBXL8 LOC105369155 ELMO3 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | MIR328 EXOC3L1 LRRC29 E2F4 FHOD1 SLC9A5 KIAA0895L PLEKHG4 LOC105369155 ELMO3 TMEM208 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | MIR328 EXOC3L1 SLC9A5 NOL3 LOC105369155 LRRC29 E2F4 FHOD1 KIAA0895L KCTD19 PLEKHG4 ELMO3 TMEM208 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | EXOC3L1 DUS2 NOL3 LOC105369155 LOC100131303 LRRC29 E2F4 LOC107984813 ELMO3 ACD DDX28 TMEM208 MIR328 TRADD PSKH1 NRN1L DPEP2 THAP11 PSMB10 ATP6V0D1 EDC4 CTRL NUTF2 KIAA0895L KCTD19 HSF4 TSNAXIP1 TPPP3 PARD6A FAM65A SLC9A5 DPEP3 C16orf70 GFOD2 FBXL8 CTCF AGRP ENKD1 LRRC36 SLC12A4 LCAT B3GNT9 ZDHHC1 CARMIL2 FHOD1 CENPT HSD11B2 PLEKHG4 C16orf86 LOC100505942 RANBP10 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |