Assay Details
Target Gene Details
Entrez Gene ID: | 80833 |
Gene Name: | apolipoprotein L3 |
Gene Aliases: |
APOLIII, CG121, CG12_1, apoL-III |
Location: |
Chr.22:36140323-36166177 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOL3 | NM_014349.2 | 4 | 1566 | NP_055164.1 |
NM_030644.1 | 5 | 1450 | NP_085147.1 | |
NM_145639.1 | 4 | 1337 | NP_663614.1 | |
NM_145640.2 | 3 | 1240 | NP_663615.1 | |
NM_145641.2 | 5 | 2749 | NP_663616.1 | |
NM_145642.2 | 4 | 2686 | NP_663617.1 | |
XM_006724324.1 | 5 | 1328 | XP_006724387.1 | |
XM_006724325.2 | 4 | 1239 | XP_006724388.1 | |
XM_017028946.1 | 4 | 1166 | XP_016884435.1 | |
XM_017028947.1 | 5 | 1279 | XP_016884436.1 | |
XM_017028948.1 | 6 | 1583 | XP_016884437.1 | |
XM_017028949.1 | 6 | 1468 | XP_016884438.1 | |
XM_017028950.1 | 5 | 2173 | XP_016884439.1 | |
XM_017028951.1 | 4 | 2060 | XP_016884440.1 | |
XM_017028952.1 | 3 | 1086 | XP_016884441.1 | |
AF070675.1 | 4 | 1406 | AAC83233.1 | |
AF305227.1 | 4 | 1193 | AAK20213.1 | |
AK025349.1 | 4 | 1163 | ||
AY014902.1 | 5 | 1450 | AAG50344.1 | |
AY014903.1 | 4 | 1337 | AAG50345.1 | |
AY014904.1 | 3 | 1240 | AAG50346.1 | |
AY014905.1 | 4 | 1566 | AAG50347.1 | |
AY014906.1 | 5 | 2749 | AAG50348.1 | |
AY014907.1 | 4 | 2686 | AAG50349.1 | |
BC042918.1 | 4 | 1142 | AAH42918.1 | |
BC050596.1 | 4 | 1197 | AAH50596.1 | |
CR456379.1 | 3 | 1060 | CAG30265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8093n54 | Chr.22:36137894 - 36187036 on Build GRCh38 | Loss | APOL3 |
esv2751964 | Chr.22:36137737 - 36183513 on Build GRCh38 | Loss | APOL3 |
esv3647677 | Chr.22:36137690 - 36164506 on Build GRCh38 | Loss | APOL3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |