Assay Details
Target Gene Details
Entrez Gene ID: | 7082 |
Gene Name: | tight junction protein 1 |
Gene Aliases: |
ZO-1 |
Location: |
Chr.15:29699367-29968919 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | ULK4P3 LOC100289656 LOC105370751 CHRFAM7A TJP1 GOLGA8R GOLGA8T WHAMMP2 LOC105370746 GOLGA8M NSMCE3 GOLGA6L7P DKFZP434L187 LOC107984752 FAM189A1 LOC100130111 GOLGA8J LOC101060569 APBA2 PDCD6IPP2 |
nsv832945 | Chr.15:29640395 - 29831518 on Build GRCh38 | Gain | TJP1 LOC100130111 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | LOC107984752 LOC100289656 FAM189A1 TJP1 LOC100130111 WHAMMP2 NSMCE3 APBA2 GOLGA6L7P PDCD6IPP2 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | ULK4P3 ULK4P2 GOLGA8H WHAMMP2 LOC105370746 GOLGA6L7P DKFZP434L187 FAM189A1 LOC100130111 GOLGA8J LOC101927579 PDCD6IPP2 LOC100289656 LOC105370751 ARHGAP11B CHRFAM7A TJP1 GOLGA8R GOLGA8T LOC100288637 NSMCE3 LOC107984752 LOC100996413 LOC101060569 APBA2 |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | ULK4P3 LOC100289656 TJP1 GOLGA8T WHAMMP2 NSMCE3 GOLGA6L7P LOC107984752 FAM189A1 LOC100130111 GOLGA8J APBA2 PDCD6IPP2 |
nsv568675 | Chr.15:29035149 - 29916451 on Build GRCh38 | Gain | FAM189A1 TJP1 LOC100130111 NSMCE3 APBA2 |
nsv832944 | Chr.15:29568858 - 29789854 on Build GRCh38 | Gain+Loss | FAM189A1 TJP1 LOC100130111 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |