Assay Details
Target Gene Details
Entrez Gene ID: | 93377 |
Gene Name: | oligodendrocytic myelin paranodal and inner loop protein |
Gene Aliases: |
HTMP10, TMEM10, TMP10 |
Location: |
Chr.10:96343216-96359365 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OPALIN | NM_001040102.2 | 2 | 542 | NP_001035191.1 |
NM_001040103.2 | 5 | 650 | NP_001035192.1 | |
NM_001284320.1 | 3 | 578 | NP_001271249.1 | |
NM_001284321.1 | 3 | 575 | NP_001271250.1 | |
NM_001284322.1 | 5 | 693 | NP_001271251.1 | |
NM_001284323.1 | 5 | 428 | NP_001271252.1 | |
NM_001284324.1 | 5 | 321 | NP_001271253.1 | |
NM_001284326.1 | 4 | 389 | NP_001271255.1 | |
NM_001284327.1 | 4 | 282 | NP_001271256.1 | |
NM_033207.4 | 4 | 611 | NP_149984.1 | |
NR_104296.1 | 4 | 670 | ||
NR_104297.1 | 4 | 597 | ||
AF367761.2 | 4 | 551 | AAK62349.1 | |
AK289835.1 | 4 | 255 | ||
AK296462.1 | 5 | 321 | ||
AK296562.1 | 3 | 194 | ||
AK309095.1 | 3 | 826 | ||
AK316090.1 | 5 | 259 | ||
AL834229.1 | 4 | 228 | CAD38907.1 | |
BC033737.1 | 4 | 228 | AAH33737.1 | |
BP349325.1 | 4 | 389 | ||
CK023662.1 | 4 | 285 | ||
DA232955.1 | 5 | 428 | ||
DA291461.1 | 3 | 238 | ||
DA351452.1 | 4 | 313 | ||
DA399407.1 | 4 | 169 | ||
DA399715.1 | 2 | 176 | ||
DA402678.1 | ||||
HY120079.1 | 5 | 337 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831950 | Chr.10:96321215 - 96485677 on Build GRCh38 | Gain | OPALIN DNTT TLL2 |
nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain | OPALIN BLNK DNTT TLL2 |