Assay Details
Target Gene Details
Entrez Gene ID: | 219285 |
Gene Name: | sterile alpha motif domain containing 9 like |
Gene Aliases: |
C7orf6, DRIF2, UEF1 |
Location: |
Chr.7:93130054-93148401 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAMD9L | NM_001303496.1 | 5 | 6799 | NP_001290425.1 |
NM_001303497.1 | 6 | 6628 | NP_001290426.1 | |
NM_001303498.1 | 5 | 6364 | NP_001290427.1 | |
NM_001303500.1 | 4 | 6232 | NP_001290429.1 | |
NM_152703.3 | 5 | 7004 | NP_689916.2 | |
XM_005250193.3 | 6 | 6436 | XP_005250250.1 | |
XM_006715890.1 | 2 | 5805 | XP_006715953.1 | |
XM_011515903.2 | 6 | 6689 | XP_011514205.1 | |
XM_017011822.1 | 4 | 6753 | XP_016867311.1 | |
XM_017011823.1 | 5 | 6509 | XP_016867312.1 | |
AB095926.1 | 5 | 6972 | ||
AI139687.1 | 1 | 166 | ||
AL834232.1 | 1 | 2512 | CAD38910.1 |